VWR®, ピペットスタンド、回転式
Assorted, 158/229×408 mm
Catalog # 613-6077
Supplier: VWR Collection
Jump to:
Product Details & Documents
This carousel pipette stand, made from ABS, can hold up to eight single-channel, four multi-channel, or a combination of both pipettes.
- Weighted base allows for easy removal or replacement of pipettes with one hand
- Pipettes are held in a vertical position to help prevent contamination
- Conveniently rotate stand head to select or store pipettes
- Quick grip handle to move pipette stand (loaded or unloaded)
- Unit is not autoclavable, but can be cleaned with soap/water or common laboratory disinfectants
A sleek, modern design enables this stand to securely hold virtually every known brand of pipette in the marketplace, making it truly 'universal'. Inserts on the upper holders can be added or removed to accommodate the fit of different of pipette widths. Moving the stand, while keeping pipettes in place, is simple with a quick grip handle allowing the user to grab and go.
Documents
VWR® Collection Selection Catalogue
Product Literature
Specifications
- Ø×H:158/229×408 mm
- Colour:Assorted