Rankem®, Solution Bottles, Plain, Tooled Neck
Solution bottle, O.D×H: 101×230 mm, 1000 ml, Clear
RANKBS-PTN-01000
: Transene
- 容量:1000 ml
- Colour:Clear
- 説明:Solution bottle, O.D×H: 101×230 mm
- Seq:03