Rankem®, Solution Bottles, Plain, Tooled Neck
Solution bottle, O.D×H: 70×140 mm, 250 ml, Clear
Catalog # RANKBS-PTN-00250
Supplier: Transene
Jump to:
Product Details & Documents
Specifications
- 容量:250 ml
- Colour:Clear
- 説明:Solution bottle, O.D×H: 70×140 mm
- Seq:01