Orders LinkContactUsLinkComponent
Rankem®, Transfer Pipettes, Class A
Transfer pipettes, tolerance: ±0,03 ml, 25 ml
Catalog # RANKPT-V-025-A
Supplier:   Transene
Rankem®, Transfer Pipettes, Class A
Catalog # RANKPT-V-025-A
Supplier:  Transene
Rankem®, Transfer Pipettes, Class A
Transfer pipettes, tolerance: ±0,03 ml, 25 ml
Price
Inquire For Price
Log in for availability
1 * 10 items
Product Details & Documents
Transfer pipettes has accuracy as per Class A (with certificate).
  • Volumetric
  • One mark
  • Borosilicate glass
Specifications
  • 容量:
    25 ml
  • 説明:
    Transfer pipettes, tolerance: ±0,03 ml
  • Seq:
    08