Orders LinkContactUsLinkComponent
試験管、縁なし、Rankem™ (ランケム)
200 mm, 25 mm
Catalog # RANKTT-WR-25200
Supplier:   Transene
試験管、縁なし、Rankem™ (ランケム)
Catalog # RANKTT-WR-25200
Supplier:  Transene
試験管、縁なし、Rankem™ (ランケム)
200 mm, 25 mm
Price
Inquire For Price
Log in for availability
1 * 100 items
Product Details & Documents
Borosilicate glass, clear, without rim.
Specifications
  • 外径:
    25 mm
  • 長さ:
    200 mm
  • Seq:
    09