Order Entry
Japan
Orders LinkContactUsLinkComponent
試験管、縁なし、Rankem™ (ランケム)

試験管、縁なし、Rankem™ (ランケム)

75 mm, 12 mm
Catalog # RANKTT-WR-1275
Supplier:   Transene
試験管、縁なし、Rankem™ (ランケム)
Catalog # RANKTT-WR-1275
Supplier:  Transene
試験管、縁なし、Rankem™ (ランケム)

75 mm, 12 mm

Price
Inquire For Price
Log in for availability
1 * 100 items
Product Details & Documents

Borosilicate glass, clear, without rim.

Specifications
  • 外径:
    12 mm
  • 長さ:
    75 mm
  • Seq:
    02