試験管、縁なし、Rankem™ (ランケム)
100 mm, 12 mm
Catalog # RANKTT-WR-12100
Supplier: Transene
Jump to:
Product Details & Documents
Borosilicate glass, clear, without rim.
Specifications
- 外径:12 mm
- 長さ:100 mm
- Seq:03