Orders LinkContactUsLinkComponent
試験管、リム付き、Rankem™(ランケム)
試験管、リム付き、Rankem™(ランケム)
100 mm, 16 mm
Catalog # RANKTT-NR-16100
Supplier:   Transene
試験管、リム付き、Rankem™(ランケム)
Catalog # RANKTT-NR-16100
Supplier:  Transene
試験管、リム付き、Rankem™(ランケム)
100 mm, 16 mm
Price
Inquire For Price
Log in for availability
1 * 100 items
Product Details & Documents
Borosilicate glass, clear, with rim.
Specifications
  • 外径:
    16 mm
  • 長さ:
    100 mm
  • Seq:
    06