Order Entry
Japan
Orders LinkContactUsLinkComponent
試験管、リム付き、Rankem™(ランケム)

試験管、リム付き、Rankem™(ランケム)

75 mm, 12 mm
Catalog # RANKTT-NR-1275
Supplier:   Transene
試験管、リム付き、Rankem™(ランケム)
Catalog # RANKTT-NR-1275
Supplier:  Transene
試験管、リム付き、Rankem™(ランケム)

75 mm, 12 mm

Price
Inquire For Price
Log in for availability
1 * 100 items
Product Details & Documents

Borosilicate glass, clear, with rim.

Specifications
  • 外径:
    12 mm
  • 長さ:
    75 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.