Orders LinkContactUsLinkComponent
細菌培養用ピペット、Rankem™(ランケム)
細菌培養用ピペット、Rankem™(ランケム)
1,1 ml, ± 0,025
Catalog # RANKPG-B-11
Supplier:   Transene
細菌培養用ピペット、Rankem™(ランケム)
Catalog # RANKPG-B-11
Supplier:  Transene
細菌培養用ピペット、Rankem™(ランケム)
1,1 ml, ± 0,025
Price
Inquire For Price
Log in for availability
1 * 10 items
Product Details & Documents
Made of grade 3.3 borosilicate glass.
  • Graduated
Specifications
  • 容量:
    1,1 ml
  • Seq:
    01
  • Tolerance:
    ± 0,025