Orders LinkContactUsLinkComponent
シリンジフィルター、RanDisc™、ナイロン
25 mm, 0,45 µm, Nylon
  RANKNY4525SF-100PB
 :   Transene
シリンジフィルター、RanDisc™、ナイロン
  RANKNY4525SF-100PB
 :  Transene
シリンジフィルター、RanDisc™、ナイロン
25 mm, 0,45 µm, Nylon
1 * 100 items
RanDisc syringe filter with nylon membrane are ready to use syringe filters which provides high flow rates at low inlet pressures. High flow rates also ensures high total throughputs i.e. filtration requirements with volumes upto 10 to 15 ml to be clarified or ultracleaned.
  • Simple, rapid and reliable ultra cleaning of small volume samples for HPLC/GC analysis
  • For rapid small volume sterilisation
  • For particle removal, ultra cleaning and purification
  • 直径:
    25 mm
  • Membrane:
    Nylon
  • 細孔サイズ:
    0,45 µm
  • Seq:
    08