Order Entry
Japan
Orders LinkContactUsLinkComponent
フラスコ、円錐形、ガラス、RankemTM

フラスコ、円錐形、ガラス、RankemTM

1000 ml, 228 mm
Catalog # RANKFC-SC-1000
Supplier:   Transene
フラスコ、円錐形、ガラス、RankemTM
Catalog # RANKFC-SC-1000
Supplier:  Transene
フラスコ、円錐形、ガラス、RankemTM

1000 ml, 228 mm

Price
Inquire For Price
Log in for availability
1 * 12 items
Product Details & Documents

Conical flasks made of borosilicate glass.

  • With screw cap
Specifications
  • 容量:
    1000 ml
  • 高さ:
    228 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.