Order Entry
Japan
Orders LinkContactUsLinkComponent
フラスコ、円錐形、ガラス、RankemTM

フラスコ、円錐形、ガラス、RankemTM

500 ml, 180 mm
Catalog # RANKFC-SC-0500
Supplier:   Transene
フラスコ、円錐形、ガラス、RankemTM
Catalog # RANKFC-SC-0500
Supplier:  Transene
フラスコ、円錐形、ガラス、RankemTM

500 ml, 180 mm

Price
Inquire For Price
Log in for availability
1 * 12 items
Product Details & Documents

Conical flasks made of borosilicate glass.

  • With screw cap
Specifications
  • 容量:
    500 ml
  • 高さ:
    180 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.