ガラスマイクロファイバーフィルタ、RankemTM
GMF sheet, Grade A, 203×254 mm
Catalog # RANKGMF-S-10
Supplier: Transene
Jump to:
Product Details & Documents
Glass microfiber filters are made of 100% borosilicate glass, and it is highly pure and extremely white in colour. Fast flow rates with high load capacity and high retention of fine particles. It can even withstand temperature up to 500 °C.
These Glass microfiber filters find application in:
- Air pollution monitoring and estimation of the suspended particles in air
- As a pre filter in highly coarse sample in HPLC analysis ideal for pre filtration to enhance sample volume of syringe filters
- Removal of precipitates in ion-pair reagents
- Clarification of reagents and solutions in spectrophotometry
- Determination of sediment in petroleum products
These are completely binder free, hence no leaching and no contamination of filtrate, biologically inert, non hygroscopic.
Specifications
- Grade:GMF sheet, Grade A
- Seq:02
- サイズ:203×254 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



