真空デシケーター、Rankem™ (ランケム)
Desiccator, 250 mm
RANKDS-TC-250-PE
: Transene
These Rankem™ desiccators are made of soft glass.
- With tubulated cover
- Feature stopper with PTFE spindle
- 内径:250 mm
- 説明:Desiccator
- Seq:03
These Rankem™ desiccators are made of soft glass.