Orders LinkContactUsLinkComponent
真空デシケーター、Rankem™ (ランケム)
Desiccator, 200 mm
  RANKDS-TC-200-PE
 :   Transene
真空デシケーター、Rankem™ (ランケム)
  RANKDS-TC-200-PE
 :  Transene
真空デシケーター、Rankem™ (ランケム)
Desiccator, 200 mm
1 * 1 items
These Rankem™ desiccators are made of soft glass.
  • With tubulated cover
  • Feature stopper with PTFE spindle
  • 内径:
    200 mm
  • 説明:
    Desiccator
  • Seq:
    02