真空デシケーター、Rankem™ (ランケム)
Desiccator, 150 mm
Catalog # RANKDS-TC-150-PE
Supplier: Transene
Jump to:
Product Details & Documents
These Rankem™ desiccators are made of soft glass.
- With tubulated cover
- Feature stopper with PTFE spindle
Specifications
- 内径:150 mm
- 説明:Desiccator
- Seq:01