Orders LinkContactUsLinkComponent
真空デシケーター、Rankem™ (ランケム)
真空デシケーター、Rankem™ (ランケム)
Desiccator, 150 mm
Catalog # RANKDS-TC-150-PE
Supplier:   Transene
真空デシケーター、Rankem™ (ランケム)
Catalog # RANKDS-TC-150-PE
Supplier:  Transene
真空デシケーター、Rankem™ (ランケム)
Desiccator, 150 mm
Product Details & Documents
These Rankem™ desiccators are made of soft glass.
  • With tubulated cover
  • Feature stopper with PTFE spindle
Specifications
  • 内径:
    150 mm
  • 説明:
    Desiccator
  • Seq:
    01