秤量ボトル、ガラス、Rankem™(ランケム)
40 ml, 60×40 mm
RANKBW-IC-40
: Transene
Weighing bottles with interchangeable stopper, made of borosilicate glass.
- Ø×H:60×40 mm
- 容量:40 ml
- Seq:05
Weighing bottles with interchangeable stopper, made of borosilicate glass.