Order Entry
Japan
Orders LinkContactUsLinkComponent
秤量ボトル、ガラス、Rankem™(ランケム)

秤量ボトル、ガラス、Rankem™(ランケム)

15 ml, 25×60 mm
Catalog # RANKBW-IC-15
Supplier:   Transene
秤量ボトル、ガラス、Rankem™(ランケム)
Catalog # RANKBW-IC-15
Supplier:  Transene
秤量ボトル、ガラス、Rankem™(ランケム)

15 ml, 25×60 mm

Price
Inquire For Price
Log in for availability
1 * 12 items
Product Details & Documents

Weighing bottles with interchangeable stopper, made of borosilicate glass.

Specifications
  • Ø×H:
    25×60 mm
  • 容量:
    15 ml
  • Seq:
    02