Order Entry
Japan
Orders LinkContactUsLinkComponent
 

秤量ボトル、ガラス、Rankem™(ランケム)

5 ml, 20×40 mm
Catalog # RANKBW-IC-05
Supplier:   Transene
秤量ボトル、ガラス、Rankem™(ランケム)
Catalog # RANKBW-IC-05
Supplier:  Transene
秤量ボトル、ガラス、Rankem™(ランケム)

5 ml, 20×40 mm

Price
Inquire For Price
Log in for availability
1 * 24 items
Product Details & Documents

Weighing bottles with interchangeable stopper, made of borosilicate glass.

Specifications
  • Ø×H:
    20×40 mm
  • 容量:
    5 ml
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.