Order Entry
Japan
Orders LinkContactUsLinkComponent
 

HPLCカラム、亜鉛

Guard cartridge, Zinc, 3,20 mm, 10 mm
  HICHZN10-10C5
   Low Manufacturing Impact
 :   HICHROM
HPLCカラム、亜鉛
  HICHZN10-10C5
 :  HICHROM
   Low Manufacturing Impact
HPLCカラム、亜鉛
HPLCカラム、亜鉛

Guard cartridge, Zinc, 3,20 mm, 10 mm

 
 
   
1 * 1 items
 
 
  • カラム形式:
    Guard cartridge
  • 内径:
    3,20 mm
  • 長さ:
    10 mm
  • Seq:
    02
  • Phase:
    Zinc
  • Cat. No.:
    HICHZN10-10C5
  • Environmental certification:
    ISO 14001:2015 (530840 UM15)
  • Environmentally Preferable:
    Low Manufacturing Impact