Avantor® Hichrom, ナットとフェルール
Valco compatible male nut
HICHHI-524
: HICHROM
Precision-machined stainless steel nuts and ferrules.
- Burr and contaminant free
- 説明:Valco compatible male nut
- Seq:23
Precision-machined stainless steel nuts and ferrules.