VWR® Basic, Overalls, with Hood
White, M
Catalog # 113-9015
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable overalls, with hood, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Hood
- Zip closure (one direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
包装: 1 piece per bag, 50 bags per case.
Documents
Specifications
- 衣服のサイズ:M
- Colour:White
- Seq:02
- Cat. No.:113-9015
- Garment application:General Purpose|Laboratory
- Weight:40 g/m²