VWR®, バイザー付きブーファントキャップ、
Blue
Catalog # 113-8909
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable caps, with peak, are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Available in white and blue
Weight: 10 g/m² (cap), 100 g/m² (peak)
包装: 100 pieces per bag, 10 bags per case.
Documents
Food contact certificate F
Additional Product Details
TDS VWR® Bouffant Cap Visor
Datasheet
VWR® Non Regulated Disposables for Production
Product Literature
Specifications
- Colour:Blue
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



