VWR®, ピークキャップ、
White
Catalog # 113-8906
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable caps, with peak, are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Available in white and blue
Weight: 40 g/m² (top), 100 g/m² (peak)
: 100 pieces per bag, 10 bags per case.
Specifications
- Colour:White