VWR®, スヌードキャップ、
Blue
Catalog # 113-8905
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable caps, with hairnet and peak, are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Total hair covering, thanks to hairnet
- Available in white and blue
Weight: 10 g/m² (hairnet), 40 g/m² (top), 100 g/m² (peak)
Packaging: 100 pieces per bag, 10 bags per case.
Documents
Food contact certificate F
Additional Product Details
TDS VWR® Snood Cap Hairnet
Datasheet
VWR® Non Regulated Disposables for Production
Product Literature
Specifications
- Colour:Blue
- Cat. No.:113-8905
- Garment application:Food



