非滅菌安全ゴーグル、VWR®
Product Details & Documents
Economical nonsterile safety goggles are designed to protect from hazards with a flexible ridge around a PVC frame.
- Two ventilation options available
- Goggles fit over most prescription glasses
- Fully adjustable headband
- Provides 99,9% UV protection
These economical, lightweight, flexible mask goggles offer unmatched comfort and protection
The indirect vent goggles feature quad vents to minimize lens fogging. Indirect vents offer protection from chemical splashes and impact.
The direct vent goggles offer direct perforations in the mask to provide maximum airflow and prevent fogging. Direct vent option provides protection from flying particles.
They can be worn over personal glasses. A broad bearing surface around the edge of the mask molds snugly to the wearer's face. The anti-fog lens coating provides superior fogging protection. The goggles meet ANSI Z87 requirements.
Frame material: PVC
Certifications: Meet ANSI Z87+ 1-2015 standards.
Specifications
- Type:Uncoated
- 説明:Direct ventilation
- Seq:02
- Lenses:Polycarbonate