Order Entry
Japan
Orders LinkContactUsLinkComponent
 

マイクロチューブラック、80箇所

VWR® 80-well microtube rack, Yellow, 80 (5×16), 1,5/2,0 ml, 225×67×28 mm
  VWRU82024-471
マイクロチューブラック、80箇所
  VWRU82024-471
マイクロチューブラック、80箇所
マイクロチューブラック、80箇所

VWR® 80-well microtube rack, Yellow, 80 (5×16), 1,5/2,0 ml, 225×67×28 mm

 
$43.43
   
1 * 5 items
 

Fraction collector plate style rack made of PP.

  • Moulded-in coordinates to aid orientation
  • Conical shaped holes (Ø 11 mm) to hold microtubes securely
  • Easily fill and transfer 80 tubes in compact arrangement (5×16 array)
 
  • For tubes:
    1,5/2,0 ml
  • 穴の数:
    80 (5×16)
  • Colour:
    Yellow
  • 説明:
    VWR® 80-well microtube rack
  • Seq:
    03
  • W×D×H:
    225×67×28 mm