Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, ブロックヒーター、アナログ

VWR®, ブロックヒーター、アナログ

Block heater, analogue, 2, RT* +5...100 °C /
75...150 °C, ±2,0 °C, 0,21 kW, 196×348×117 mm, 2,6 kg
Catalog # 460-0355
Supplier:   VWR Collection
VWR®, ブロックヒーター、アナログ
Catalog # 460-0355
Supplier:  VWR Collection
VWR®, ブロックヒーター、アナログ
VWR®, ブロックヒーター、アナログ

Block heater, analogue, 2, RT* +5...100 °C /
75...150 °C, ±2,0 °C, 0,21 kW, 196×348×117 mm, 2,6 kg

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Multi-purpose, high wattage, constant temperature, VWR® analogue block heaters are economical, versatile and compact.

  • Compact design with small footprint
  • Temperature range, low: From ambient +5 to 100 °C, high: 75 to 150 °C
  • Multi-purpose device for a wide range of applications
  • Exceptional temperature uniformity
  • Accommodates VWR® interchangeable, modular heating blocks for tubes from 0,2 up to 50 ml capacity
  • Can be used at 18 to 33 °C, with 20 to 80% relative humidity, non condensing

These multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a variety of other laboratory procedures. Two separate temperature adjustment knobs provide low and high temperature control. Low range knob adjusts from above ambient to 100 °C, and high range knob adjusts from 75 to 150 °C. Each unit requires minimum bench space.

Each model accepts separate interchangeable, modular heating blocks, which accommodates various sample containers such as microtubes, centrifuge tubes, vials, microplates and PCR strips or tubes. Blocks include a thermometer well, and feature close 'tube and block' contact for maximum heat retention.

Certifications: TUV listed, RoHS compliant.

注文情報: Supplied without heating blocks, please order these separately.

注意: To avoid a possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed for use as a dry bath/incubator only.
* RT = Ambient

Specifications
  • Heating capacity:
    0,21 kW
  • No. of blocks:
    2
  • 温度範囲:
    RT* +5...100 °C /
    75...150 °C
  • Weight:
    2,6 kg
  • Seq:
    002
  • W×D×H:
    196×348×117 mm
  • Temp. stability:
    ±2,0 °C
  • Type:
    Block heater, analogue
Accessories

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3282
    Single block
    Ø 19,7 mm
    19 mm vials
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3283
    Single block
    Ø 21,7 mm
    21 mm vials
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3215
    Single block
    Ø 13,9 mm
    12/13 mm round bottom tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3216
    Single block
    Ø 13,9 mm
    12/13 mm round bottom tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3217
    Single block
    Ø 17,5 mm
    15/16 mm round bottom tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    穴の数
    Availability
    Price
  • 460-3248
    Single block, microtube combination 0,5 ml, 1,5 ml, 2,0 ml
    Ø 7,9 / Ø 11,1 / Ø 11,5 mm
    6/10/5
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3211
    Single block
    Ø 7,9 mm
    0,5 ml PCR tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3212
    Single block
    Ø 11,5 mm
    1,5 ml Eppendorf microtubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3234
    Single block
    Ø 6,3 mm
    Round bottomed tubes, 6 mm Ø
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3235
    Single block
    Ø 11,1 mm
    1,5 ml VWR microtubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3245
    Single block
    Ø 11,5 mm
    2,0 ml VWR/Eppendorf microtubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3246
    Single block
    Ø 10,9 mm
    2,0 ml Corning microtubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3221
    Single block
    Ø 17,1 mm
    15 ml conical tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3223
    Single block
    Ø 29,0 mm
    50 ml conical tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    W×D×H
    Availability
    Price
  • 460-3239
    Double block
    - mm
    95×152×57 mm
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3238
    Double block
    - mm
    96- or 384-well plate
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3224
    Single block, 10×8-PCR tube strips
    Ø 6,4 mm
    0,2 ml tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3225
    Single block, 64× individual PCR tubes
    Ø 6,4 mm
    0,2 ml tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3226
    Double block, 1×96-well PCR plate
    Ø 6,4 mm
    0,2 ml tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    穴の数
    Availability
    Price
  • 460-3237
    Single block, 2×6 cuvettes (12,5 mm)
    - mm
    -
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    W×D×H
    Availability
    Price
  • 460-3236
    Single block
    - mm
    95×76×51 mm
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3279
    Single block
    Ø 12,7 mm
    12 mm vials
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3280
    Single block
    Ø 15,8 mm
    15 mm vials
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3281
    Single block
    Ø 17,8 mm
    17 mm vials
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3285
    Single block
    Ø 25,8 mm
    25 mm vials
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3284
    Single block
    Ø 23,8 mm
    23 mm vials
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3286
    Single block
    Ø 28,8 mm
    28 mm vials
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3287
    Single block
    Ø 16,4 mm
    16 mm vials
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3213
    Single block
    Ø 8,3 mm
    6 mm round bottom tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3214
    Single block
    Ø 10,7 mm
    10 mm round bottom tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3218
    Single block
    Ø 21,0 mm
    20 mm round bottom tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3219
    Single block
    Ø 26,2 mm
    25 mm round bottom tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3222
    Single block
    Ø 35,0 mm
    35 mm round bottom tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3243
    Single block
    Ø 19,1 mm
    17/18 mm round bottom tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • VWRU14216-212
    Single block
    Ø 19,1 mm
    17/18 mm round bottom tubes
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    穴の数
    Availability
    Price
  • 460-3220
    Single block, test tube combination 6 mm, 12/13 mm, 25 mm
    Ø 8,3 / Ø 13,9 / Ø 26,2 mm
    6/5/3
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Well size
    穴の数
    Availability
    Price
  • 460-3247
    Single block, centrifuge tube combination 1,5 ml, 15 ml, 50 ml
    Ø 11,1 / Ø 17,1 / Ø 29,0 mm
    4/3/2
    Log in for availability
    1 * 1 items