VWR®, 秤量ボトル、ガラス
80 ml, 80×30 mm, 80/12
Borosilicate glass 3,3, low form.
- Interchangeable knobbed lid with ground joint
- White labelling area
- Resistant to chemicals and thermal shock
- Ø×H:80×30 mm
- NS:80/12
- 容量:80 ml
- Seq:06