Orders LinkContactUsLinkComponent
VWR®, 秤量ボトル、ガラス
30 ml, 50×30 mm, 50/12
Catalog # 611-3820
Supplier:   VWR Collection
VWR®, 秤量ボトル、ガラス
Catalog # 611-3820
Supplier:  VWR Collection
VWR®, 秤量ボトル、ガラス
VWR®, 秤量ボトル、ガラス
30 ml, 50×30 mm, 50/12
Product Details & Documents
Borosilicate glass 3,3, low form.
  • Interchangeable knobbed lid with ground joint
  • White labelling area
  • Resistant to chemicals and thermal shock
Specifications
  • Ø×H:
    50×30 mm
  • NS:
    50/12
  • 容量:
    30 ml
  • Seq:
    04