VWR®, 秤量ボトル、ガラス
15 ml, 40×25 mm, 40/12
Borosilicate glass 3,3, low form.
- Interchangeable knobbed lid with ground joint
- White labelling area
- Resistant to chemicals and thermal shock
- Ø×H:40×25 mm
- NS:40/12
- 容量:15 ml
- Seq:03
Borosilicate glass 3,3, low form.