VWR®, 秤量ボトル、ガラス
6 ml, 25×25 mm, 24/12
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate glass 3,3, low form.
- Interchangeable knobbed lid with ground joint
- White labelling area
- Resistant to chemicals and thermal shock
Specifications
- Ø×H:25×25 mm
- NS:24/12
- 容量:6 ml
- Seq:01