Orders LinkContactUsLinkComponent
VWR®, ビーカー、低形状、PP
VWR®, ビーカー、低形状、PP
5000 ml
Catalog # 213-0496
   Sustainable Packaging
Supplier:   VWR Collection
VWR®, ビーカー、低形状、PP
Catalog # 213-0496
Supplier:  VWR Collection
   Sustainable Packaging
VWR®, ビーカー、低形状、PP
VWR®, ビーカー、低形状、PP
5000 ml
Product Details & Documents
PP, translucent.
  • Printed graduations
  • Can be stacked when empty to save space while storing
  • Withstand temperatures of 0 to 121 °C
  • Resin meets US FDA 21 CFR requirements
Ideal for general laboratory use. With spout and exceptionally 'easy to read' printed graduations.
Specifications
  • 容量:
    5000 ml
  • Seq:
    09
  • Cat. No.:
    213-0496
  • Environmentally Preferable:
    Sustainable Packaging