Orders LinkContactUsLinkComponent
VWR®, ビーカー、低形状、PP
VWR®, ビーカー、低形状、PP
1000 ml
Catalog # 213-0493
   Sustainable Packaging
Supplier:   VWR Collection
VWR®, ビーカー、低形状、PP
Catalog # 213-0493
Supplier:  VWR Collection
   Sustainable Packaging
VWR®, ビーカー、低形状、PP
VWR®, ビーカー、低形状、PP
1000 ml
Price
Inquire For Price
Log in for availability
1 * 3 items
Product Details & Documents
PP, translucent.
  • Printed graduations
  • Can be stacked when empty to save space while storing
  • Withstand temperatures of 0 to 121 °C
  • Resin meets US FDA 21 CFR requirements
Ideal for general laboratory use. With spout and exceptionally 'easy to read' printed graduations.
Specifications
  • 容量:
    1000 ml
  • Seq:
    06
  • Cat. No.:
    213-0493
  • Environmentally Preferable:
    Sustainable Packaging