VWR®, ビーカー、低形状、PP
500 ml
PP, translucent.
- Printed graduations
- Can be stacked when empty to save space while storing
- Withstand temperatures of 0 to 121 °C
- Resin meets US FDA 21 CFR requirements
Ideal for general laboratory use. With spout and exceptionally 'easy to read' printed graduations.
- 容量:500 ml
- Seq:05
- Cat. No.:213-0492
- Environmentally Preferable:Sustainable Packaging
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



