Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, ビーカー、低形状、PP

VWR®, ビーカー、低形状、PP

100 ml
Catalog # 213-0490
   Sustainable Packaging
Supplier:   VWR Collection
VWR®, ビーカー、低形状、PP
Catalog # 213-0490
Supplier:  VWR Collection
   Sustainable Packaging
VWR®, ビーカー、低形状、PP
VWR®, ビーカー、低形状、PP

100 ml

Price
Inquire For Price
Log in for availability
1 * 6 items
Product Details & Documents

PP, translucent.

  • Printed graduations
  • Can be stacked when empty to save space while storing
  • Withstand temperatures of 0 to 121 °C
  • Resin meets US FDA 21 CFR requirements

Ideal for general laboratory use. With spout and exceptionally 'easy to read' printed graduations.

Specifications
  • 容量:
    100 ml
  • Seq:
    03
  • Cat. No.:
    213-0490
  • Environmentally Preferable:
    Sustainable Packaging
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.