VWR®, ビーカー、低形状、PP
25 ml
Catalog # 213-0511
Supplier: VWR Collection
Jump to:
Product Details & Documents
PP, translucent.
- Printed graduations
- Can be stacked when empty to save space while storing
- Withstand temperatures of 0 to 121 °C
- Resin meets US FDA 21 CFR requirements
Ideal for general laboratory use. With spout and exceptionally 'easy to read' printed graduations.
Documents
Specifications
- 容量:25 ml
- Seq:01
- Cat. No.:213-0511
- Environmentally Preferable:Sustainable Packaging