VWR®, ガラスビーズ
4,0 mm
Catalog # 201-1254
Supplier: VWR Collection
Jump to:
Product Details & Documents
Soda-lime glass, solid.
- Not suitable for heat applications
- Available in sizes from 1 to 10 mm
Can be used for many different applications in the laboratory. In microbiology laboratories they are used for the production of single cell suspensions or for plating. In chemical laboratories they are used as filling material for distillation columns. They can also be used to clean round bottom vessels.
Documents
CoC VWR Glass beads.pdf
Conformity declaration
VWR® Glassware Solutions
Product Literature
Specifications
- 直径:4,0 mm
- Seq:04