Orders LinkContactUsLinkComponent
VWR®, ガラスビーズ
2,0 mm
  201-1252
VWR®, ガラスビーズ
  201-1252
VWR®, ガラスビーズ
VWR®, ガラスビーズ
2,0 mm
1 * 500 g
Soda-lime glass, solid.
  • Not suitable for heat applications
  • Available in sizes from 1 to 10 mm
Can be used for many different applications in the laboratory. In microbiology laboratories they are used for the production of single cell suspensions or for plating. In chemical laboratories they are used as filling material for distillation columns. They can also be used to clean round bottom vessels.
  • 直径:
    2,0 mm
  • Seq:
    02