VWR®, ワークステーション用ディスペンサー
Workstation dispensing bin, 4 compartments, 278×158×217 mm
Catalog # 216-2005
Supplier: VWR Collection
Jump to:
Product Details & Documents
Crystal-clear bins for small inventory items, like pipette tips, tubes, fittings, finger cots and other small bulk items.
- Flip-top covers on generous openings, make filling and dispensing easy and keep products dust-free
- Four sizes available to cater to a wide variety of storage needs
- Includes non skid rubber feet
Documents
VWR® Collection Selection Catalogue
Product Literature
Specifications
- 説明:Workstation dispensing bin, 4 compartments
- Seq:04
- W×D×H:278×158×217 mm