Orders LinkContactUsLinkComponent
VWR®, ワークステーション用ディスペンサー
Workstation dispensing bin, 2 compartments, 139×160×368 mm
Catalog # 216-2003
Supplier:   VWR Collection
VWR®, ワークステーション用ディスペンサー
Catalog # 216-2003
Supplier:  VWR Collection
VWR®, ワークステーション用ディスペンサー
VWR®, ワークステーション用ディスペンサー
Workstation dispensing bin, 2 compartments, 139×160×368 mm
Product Details & Documents
Crystal-clear bins for small inventory items, like pipette tips, tubes, fittings, finger cots and other small bulk items.
  • Flip-top covers on generous openings, make filling and dispensing easy and keep products dust-free
  • Four sizes available to cater to a wide variety of storage needs
  • Includes non skid rubber feet
Specifications
  • 説明:
    Workstation dispensing bin, 2 compartments
  • Seq:
    02
  • W×D×H:
    139×160×368 mm