Order Entry
Japan
Orders LinkContactUsLinkComponent
 

スプーンヘラ, 磁器

165 mm, 0,5 ml
Catalog # VWRU89038-206
Supplier:   VWR Collection
スプーンヘラ, 磁器
Catalog # VWRU89038-206
Supplier:  VWR Collection
スプーンヘラ, 磁器
スプーンヘラ, 磁器

165 mm, 0,5 ml

Price
$76.88
Log in for availability
1 * 1 items
Product Details & Documents

Chemical-porcelain spatulas feature a spatula on one end and a spoon on the other.

  • High chemical resistance
  • Glazed for added protection
Specifications
  • 容量:
    0,5 ml
  • 長さ:
    165 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.