Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, インクジェットプリンタスライド

VWR®, インクジェットプリンタスライド

Inkjet printer coloured-frosted slides on 20 mm, Yellow, Glass, 1,0 (-0,05/+0,1) mm, 76×25 mm
Catalog # 631-1182
Supplier:   VWR Collection
VWR®, インクジェットプリンタスライド
Catalog # 631-1182
Supplier:  VWR Collection
VWR®, インクジェットプリンタスライド
VWR®, インクジェットプリンタスライド

Inkjet printer coloured-frosted slides on 20 mm, Yellow, Glass, 1,0 (-0,05/+0,1) mm, 76×25 mm

Price
Inquire For Price
Log in for availability
1 * 2.500 items
Product Details & Documents

Microscopy slides are designed with colour-frosted ends for H&E stain on frozen sections in histopathology examination.

  • Shelf life of 60 months
  • Pre-cleaned, without mark, print or contamination risks
  • Marking with conventional labelling system, pencil, marker pens and inkjet printers

Specially produced for all inkjet printers.

 : 50 pieces per box. 50 boxes per case.

 : Fragile items should be stored in dry and proper laboratory temperature conditions.

Specifications
  • Colour:
    Yellow
  • 説明:
    Inkjet printer coloured-frosted slides on 20 mm
  • 長さ×幅:
    76×25 mm
  • Material:
    Glass
  • 厚さ:
    1,0 (-0,05/+0,1) mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.