Orders LinkContactUsLinkComponent
VWR®, カラーフロスト顕微鏡スライド
Colour-frosted microscope slides, Green, 1,0 (-0,05/+0,1) mm, 76×25 mm
Catalog # 631-1172
Supplier:   VWR Collection
VWR®, カラーフロスト顕微鏡スライド
Catalog # 631-1172
Supplier:  VWR Collection
VWR®, カラーフロスト顕微鏡スライド
VWR®, カラーフロスト顕微鏡スライド
Colour-frosted microscope slides, Green, 1,0 (-0,05/+0,1) mm, 76×25 mm
Price
Inquire For Price
Log in for availability
1 * 2.500 items
Product Details & Documents
Microscope slides recommended for haematoxylin and eosin (H&E) stain and hybridisation on frozen sections in histopathology examination.
  • Smooth edges for operational safety
  • Sticky surface for stronger adhesion than with poly-l-lysine
  • Marking with conventional labelling system, pencils, marker pens and thermal transfer printers
Recommended for hematoxylin and eosin (H&E) stain, immunohistochemistry (IHC) and hybridisation on frozen sections in histopathology examination.
 : 50 pieces per box. 50 boxes per case.
 : To be stored in dry and proper laboratory temperature conditions − shelf life of 60 months.
Specifications
  • 幅×奥行き:
    76×25 mm
  • Colour:
    Green
  • 説明:
    Colour-frosted microscope slides
  • 厚さ:
    1,0 (-0,05/+0,1) mm
  • Cat. No.:
    631-1172