VWR®, ビーカー、ローフォーム、PMP
1000 ml, 20 ml
Catalog # VWRU89044-518
Supplier: VWR Collection
Jump to:
Product Details & Documents
PMP, crystal clear, graduated.
- Excellent chemical resistance
- Autoclavable
- Withstand a temperature of up to 170 °C for short periods
With large spout and easy to read blue graduations.
Certifications: Conform to ISO 7056 1981 (E) and BS 5404 Part 1.
Specifications
- 目盛り間隔:20 ml
- 容量:1000 ml
- Seq:04