VWR®, VisiCam®用カメラアダプター
C-mount adapter for 1/3” sensor
Catalog # 630-2720
Supplier: VWR Collection
Jump to:
Product Details & Documents
Documents
VWR® for Microscopy
Product Literature
Specifications
- 説明:C-mount adapter for 1/3” sensor
- Seq:217