VWR®, ロータリーベーンポンプ用アクセサリー
Separator on the suction side of the rotary vane pump - separates liquid and particles from the inlet gas (can be installed directly on the inlet flange)
Catalog # 181-7003
Supplier: VWR Collection
Jump to:
Product Details & Documents
Documents
181-7002 181-7002_b 181-7007
Images 360
Specifications
- 説明:Separator on the suction side of the rotary vane pump - separates liquid and particles from the inlet gas (can be installed directly on the inlet flange)
- Seq:02