Orders LinkContactUsLinkComponent
VWR®, ロータリーベーンポンプ用アクセサリー
Oil mist separator on the pressure side of the rotary vane pump - catches the small amounts of oil in the exhaust gas of the pump (can be be installed directly on the exhaust flange)
  181-7002
VWR®, ロータリーベーンポンプ用アクセサリー
  181-7002
VWR®, ロータリーベーンポンプ用アクセサリー
VWR®, ロータリーベーンポンプ用アクセサリー
Oil mist separator on the pressure side of the rotary vane pump - catches the small amounts of oil in the exhaust gas of the pump (can be be installed directly on the exhaust flange)
1 * 1 items
  • 説明:
    Oil mist separator on the pressure side of the rotary vane pump - catches the small amounts of oil in the exhaust gas of the pump (can be be installed directly on the exhaust flange)
  • Seq:
    01