VWR®, ロータリーベーンポンプ用アクセサリー
Oil mist separator on the pressure side of the rotary vane pump - catches the small amounts of oil in the exhaust gas of the pump (can be be installed directly on the exhaust flange)
181-7002
- 説明:Oil mist separator on the pressure side of the rotary vane pump - catches the small amounts of oil in the exhaust gas of the pump (can be be installed directly on the exhaust flange)
- Seq:01