VWR®, デジタルミニブロックヒーター
Mini block heater with heated lid***, 1, 0,075 kW, Ambient +5...100 °C, ±0,2 °C, ±0,3 °C
460-0337
These mini block heaters are ideal for applications that require accurate, repeatable results. These personal-sized units take up minimal bench space. Choice of model with or without heated lid. Units are ideal for immunoassays, ELISA, LAMP assays, enzyme reactions, denaturations and blood banking.
- LED display for temperature and time
- Temperature range from ambient 5 to 100 °C
- Temperature accuracy: ±0,5 °C (models with heated lid: ±0,3 °C)
- Microprocessor controlled with temperature calibration via external thermometer or temperature device
- Timer range: 1 second to 160 hours, or continuous mode
- Heated lid helps to reduce condensation within sample tube
- Accommodates interchangeable mini heating blocks for tubes from 0,2 up to 50 ml capacity
Accessories information: Blocks for 15 and 50 ml tubes (460-0342 and 462-0343) cannot be used with the mini block heater with heated lid.
Ordering information: Heating blocks are not included, please order these separately.
Caution: ** EU/CH-plug *** UK-plug
- Heating capacity:0,075 kW
- No. of blocks:1
- 温度範囲:Ambient +5...100 °C
- Seq:004
- Accuracy:±0,3 °C
- Temp. stability:±0,2 °C
- Type:Mini block heater with heated lid***
- Cat. No.:460-0337
- W×D×H:127×160×123 mm
- Weight:1,3 kg



